Order Entry
United States
ContactUsLinkComponent
Random Primer 6, New England Biolabs
Random Primer 6, New England Biolabs
Catalog # 101229-748
Small Business Enterprise
CAS Number:  
undefined
Random Primer 6, New England Biolabs
Catalog # 101229-748
Supplier Number:  S1230S
Small Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Application:
  • Description:
    Random Primer 6
  • Size:
    1.0 A260 units
  • Environmentally Preferable:
  • Cat. No.:
    101229-748
  • Supplier no.:
    S1230S

Specifications

About this item

This mixture of random hexanucleotides is used to prime DNA synthesis in vitro along multiple sites of denatured template DNA

The newly synthesized complementary DNA is "oligo-labelled" by substituting any radiolabelled nucleotide for the appropriate nonradioactive nucleotide in the reaction mixture. Use of synthetic d(N6) primer ensures the presence of virtually all sequence combination of hexamer primers which results in equally labelled DNA of high specific activity. Oligolabelling by this method generates probes which can be used to screen gene libraries, probe Southern and Northern blots, and for in situ hybridizations.

This primer-template complex is a suitable substrate for DNA Polymerase I (Klenow Fragment).