ContactUsLinkComponent
QUICK-LOAD® DNA Ladder, 2-log, New England Biolabs
QUICK-LOAD® DNA Ladder, 2-log, New England Biolabs
Catalog # 101227-138
Small Business Enterprise
CAS Number:  
undefined
QUICK-LOAD® DNA Ladder, 2-log, New England Biolabs
Catalog # 101227-138
Supplier Number:  N0469S
Small Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Quick-Load 2-log DNA Ladder
  • Size:
    250 gel lanes
  • Environmentally Preferable:
  • Cat. No.:
    101227-138
  • Supplier no.:
    N0469S
Specifications
About this item
Quick-Load® 2-Log DNA Ladder is a pre-mixed, ready-to-load molecular weight marker containing bromophenol blue as a tracking dye
The DNA Ladder consists of proprietary plasmids which are digested to completion with appropriate restriction enzymes to yield 19 bands suitable for use as molecular weight standards for agarose gel electrophoresis. This digested DNA includes fragments ranging from 100 bp to 10 kb. The 0.5, 1.0 and 3.0 kb bands have increased intensity to serve as reference bands.