NEBNEXT® Small RNA Library Prep Set for Illumina (Multiplex Compatible), New England Biolabs
Supplier: New England Biolabs (NEB)
Certificates
About this item
The novel NEBNext® Small RNA workflow has been optimized to minimize adaptor-dimers while producing high-yield, high-diversity libraries
The NEBNext® Small RNA Library Prep Set for Illumina® (Multiplex Compatible) enables use with your own barcode system. The kit contains adaptors but does not include multiplex primers.
Specifications
- Application:
- Environmentally Preferable: