ContactUsLinkComponent
Lambda DNA-BstE II Digest, New England Biolabs
Lambda DNA-BstE II Digest, New England Biolabs
Catalog # 101228-444
Small Business Enterprise
CAS Number:  
undefined
Lambda DNA-BstE II Digest, New England Biolabs
Catalog # 101228-444
Supplier Number:  N3014S
Small Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Lambda DNA-BstE II Digest
  • Size:
    150 gel lanes
  • Environmentally Preferable:
  • Cat. No.:
    101228-444
  • Supplier no.:
    N3014S
Specifications
About this item
The BstEII digest of lambda DNA (cI857 ind 1 Sam 7) yields 14 fragments suitable for use as molecular weight standards for agarose gel electrophoresis
The approximate mass of DNA in each of the bands is provided (assuming a 1.0 μg load) for approximating the mass of DNA in comparably intense samples of similar size. Comes supplied with 1 vial of Gel Loading Dye, Purple (6X), no SDS.