Order Entry
United States
ContactUsLinkComponent
Anti-RELM-beta Rat Monoclonal Antibody [clone: MRB 46L]
Anti-RELM-beta Rat Monoclonal Antibody [clone: MRB 46L]
Supplier:  Adipogen
Certificates
Anti-RELM-beta Rat Monoclonal Antibody [clone: MRB 46L]
Supplier:  Adipogen
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Catalog No:
  • 102514-008
  • 102514-010
  • Antigen Symbol:
  • RELMB
  • RELMB
  • Antigen Name:
  • Resistin-like beta
  • Resistin-like beta
  • Antigen Synonyms:
  • HXCP2|FIZZ2|Resistin-like Molecule-beta
  • HXCP2|FIZZ2|Resistin-like Molecule-beta
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • MRB 46L
  • MRB 46L
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • Q99P86
  • Q99P86
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • Western Blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant mouse RELM-beta.
  • Recombinant mouse RELM-beta.
  • Form:
  • Liquid
  • Liquid
  • Concentration:
  • 1mg/ml
  • 1mg/ml
  • Storage Buffer:
  • 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives.
  • 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives.
  • Storage Temperature:
  • Short term: +4 °C, Long Term: –20 °C
  • Short term: +4 °C, Long Term: –20 °C
  • Shipping Temperature:
  • -20° C
  • -20° C

Specifications