To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Resistin
- Antigen Symbol:RETN
- Clonality:Monoclonal
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Rat
- Western Blot:Yes
- Size:50 µg
- Environmentally Preferable:
- Form:Liquid. In PBS, pH 7.4. Contains no preservatives.
- Antigen Synonyms:Cysteine-rich secreted protein FIZZ3|Adipose tissue-specific secretory factor|C/EBP-epsilon-regulated myeloid-specific secreted cysteine-|RSN|ADSF
- Concentration:1mg/ml
- Cat. No.:89154-470
- Supplier no.:804-525-C050
Specifications
About this item
Host: Mouse, Isotype: IgG1
- For ELISA, Flow Cytometry, Western Blot
Recognizes both natural and recombinant rat resistin. Detects a band of ~10kDa by Western blot. Does not cross-react with human and mouse resistin.
Type: Primary
Antigen: RETN
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat