To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Tumor Necrosis Factor (Ligand) Superfamily, Member 10
- Antigen Symbol:TNFSF10
- Clonality:Polyclonal
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western Blot:Yes
- Size:100 µL
- Environmentally Preferable:
- Form:Liquid. Contains 0.02% sodium azide.
- Antigen Synonyms:Tumor necrosis factor ligand superfamily member 10|Apo-2 ligand|Apo-2L|Protein TRAIL|TNF-related apoptosis-inducing ligand|CD253
- Cat. No.:89146-022
- Supplier no.:210-732-R100
Specifications
About this item
Host: Rabbit
- For Immunocytochemistry, Immunohistochemistry, Western Blot
Recognizes human soluble TRAIL. Does not cross-react with mouse TRAIL.
Type: Primary
Antigen: TNF-related Apoptosis-inducing Ligand
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human