Human Recombinant SGK-1 (truncated NT 60 aa) (GST-tag) (from Insect Cells (Sf9)), GST Tag
Catalog # 89142-020
Supplier: Avantor
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Protein/Peptide Type:Recombinant
- Source:Insect cells (Sf9)
- Species:Human
- Size:5 µg
- Environmentally Preferable:
- Tag sequence:GST Tag
- Form:Liquid
- Protein/Peptide Name:SGK-1 (truncated NT 60 aa) (GST-tag)
- Purity:>90% (SDS-PAGE)
- Formulation:In 50mM TRIS-HCl, pH 7.5, containing 150mM sodium chloride, 0.25mM DTT, 0.1mM EGTA, 0.1mM EDTA, 0.1mM PMSF, and 25% glycerol.
- Purification:Purified by multi-step chromatography.
- Cat. No.:89142-020
- Supplier no.:PPK-459-Z
Specifications
About this item
Active recombinant human SGK-1 (truncated NT 60 aa) produced in Sf9 cells using an N-terminal GST-tag.