Order Entry
United States
ContactUsLinkComponent
 
Hard-Shell® 96-Well Semi-Skirted PCR Plates, High-Profile, Bio-Rad
  101647-160
 :  Bio-Rad
undefined
Hard-Shell® 96-Well Semi-Skirted PCR Plates, High-Profile, Bio-Rad
  101647-160
 :  Bio-Rad
 :  HSS9601
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Frame color:
    Clear
  • No. of wells:
    96
  • Skirt:
    Semi-Skirted
  • Well Color:
    Clear
  • Well volume:
    0,35 ml
  • Description:
    PCR Plates
  • Environmentally Preferable:
  • Cat. No.:
    101647-160
  • Barcoded:
    No
  • Supplier no.:
    HSS9601

 

 

Hard-Shell 96-Well Semi-Skirted PCR Plates are designed to withstand the stresses of thermal cycling and robotic handling

  • Reaction volumes of 5–125 µL (350 µL maximum)
  • Black lettering for easy well identification
  • Free of DNase, RNase and human DNA
  • White-well option for increased fluorescent signal strength
  • Color-coded skirt option with clear wells
  • Low-cost, user-readable bar code option for convenient database tracking

The raised rims around each well allow tight sealing using different procedures including pressure-, adhesive-, and heat-sealing methods. The high-profile (20.75 mm) wells fit most thermal cyclers, real-time PCR detection systems, and DNA sequencers.

The patented two-component design provides stability and flatness, allowing precise positioning for automation. The skirt and deck are molded from a rigid thermostable polymer to eliminate warping and shrinkage, and the thin-wall wells are molded of virgin polypropylene with low DNA binding. The half-height skirt provides improved robotic handling and a labeling surface.

Compatible with: CFX96™, 1000-Series, DNA Engine Opticon® Series, Chromo4™, PTC-100®, DNA Engine® Series, Applied Biosystems standard systems (7300, 7500, 9700, 2070, Veriti, etc.), Applied Biosystems Sanger sequencers (3700, 3730, etc.), Eppendorf Mastercyclers.