To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Secondary
- Antigen Name:Rabbit Whole Serum
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- Isotype:IgG
- Reactivity:Rabbit
- Environmentally Preferable:
- Storage Buffer:0.1% sodium Azide
- Storage Temperature:Keep as concentrated solution, aliquot and store at 4°C.
- Cat. No.:89366-186
- Supplier no.:GTX77172
This antiserum forms multiple arcs against normal rabbit serum by immunoelectrophoresis. Goats were immunized with pooled normal rabbit serum in Freund's adjuvant.
Type: Secondary
Antigen: Rabbit Whole Serum
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit (Lapine)