Order Entry
United States
ContactUsLinkComponent
Anti-PRF1 Mouse Monoclonal Antibody [clone: CE2.10]
Anti-PRF1 Mouse Monoclonal Antibody [clone: CE2.10]
Catalog # 89360-146
Supplier:  Genetex
Anti-PRF1 Mouse Monoclonal Antibody [clone: CE2.10]
Catalog # 89360-146
Supplier:  Genetex
Supplier Number:  GTX16079
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Perforin
  • Antigen Symbol:
    PRF1
  • Clonality:
    Monoclonal
  • Clone:
    CE2.10
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    50 μg
  • Environmentally Preferable:
  • Gene ID:
    18646
  • Antigen Synonyms:
    lymphocyte pore-forming protein|cytolysin|lymphocyte pore forming protein|perforin-1
  • Storage Buffer:
    Preservative: 0.02% Sodium Azide; Constituents: PBS
  • Storage Temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Concentration:
    0.5 mg/ml
  • Immunogen:
    Recombinant fragment, corresponding to amino acids 98 - 534 of Mouse Perforin.
  • Tested Applications:
    IP, WB
  • Purification:
    >95% by SDS-PAGE
  • Cat. No.:
    89360-146
  • Supplier no.:
    GTX16079

Specifications

About this item

Perforin is one of the major cytolytic proteins of cytolytic granules. One of the main pathways of lymphocyte mediated cytolysis entails the secretion onto target membranes of cytolytic granules contained in cytolytic effector lymphocytes of T-cell or NK-cell type. Perforin is a cytolytic mediator and is stored in and released by cytoplasmic granules. Perforin (PRF) is involved in the killing mediated by cytotoxic lymphocytes which represents an important mechanism in the immune defense against tumors and virus infections. Human PRF is a 555 amino acid protein with a 21 amino acid signal peptide. It has a molecular weight of 70 to 75 kD. PRF is a pore forming protein with a mechanism of transmembrane channel formation similar to C9 and homology between perforin and C9 has been demonstrated at their respective functionally conserved regions. Studies show that perforin is expressed only in killer cell lines and not in helper T lymphocytes or other tumor cells tested.

IP: Use at an assay dependent dilution on human and mouse samples. WB: Use at a concentration of 1 ?g/ml. Detects a band of approximately 66 kDa (predicted molecular weight: 66 kDa) from the mouse T cell clone B6.1. The antibody does not detect human perforin by Western blot. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PRF1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human