Order Entry
United States
ContactUsLinkComponent
Anti-IgG2a Mouse Monoclonal Antibody
Anti-IgG2a Mouse Monoclonal Antibody
Catalog # 89352-776
Supplier:  Genetex
Anti-IgG2a Mouse Monoclonal Antibody
Catalog # 89352-776
Supplier:  Genetex
Supplier Number:  GTX35020
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Control
  • Antigen Name:
    Immunoglobulin G2 alpha
  • Antigen Symbol:
    IgG2a
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Mouse
  • Environmentally Preferable:
  • Storage Buffer:
    PBS + 0.1% NaN3
  • Storage Temperature:
    Store at 2-8°C
  • Tested Applications:
    Fluorescence-Activated Cell Sorting
  • Cat. No.:
    89352-776
  • Supplier no.:
    GTX35020

Specifications

About this item

Inclusion of an appropriate isotype control is essential for the proper interpretation of experimental results. Because isotype control antibodies have no relevant specificity, they enable the researcher to distinguish non-specific "background" binding from antigen-specific antibody binding. Non-specific binding of primary antibodies may occur due to either binding of antibody to Fc receptors or non-specific interactions with cellular proteins, lipids, or carbohydrates. Additionally, autofluorescence or endogenous enzyme activity in target cells may give a false positive that could be misinterpreted as antibody binding. These factors vary by target cell type and the isotype of the primary antibody. Therefore, the primary antibody used should be matched to an isotype control of identical species and isotype (including both heavy and light chains).

Type: Control
Antigen: IgG2a
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse