Ethyl 3-aminocrotonate 98.5%
: Ethyl 3-aminobut-2-enoate
:7318-00-5
:626-34-6
:C₆H₁₁NO₂
:MFCD00008073
:1.022 g/cm³
:129.16 g/mol
:33…35 °C
:97 °C
:Ambient
:230-782-0
:3263
:8 III
:GHS05
- Environmentally Preferable:
- Appearance:Light yellow Low melting solid or liquid
- Infrared spectrum:Conforms
- GC:≥98.0 %
- Titration with HClO4:≥98.0 %
- Sulfate (SO4):≤0.005 %
- Ammonium (NH4):≤0.003 %
- Iron (Fe):≤2 ppm
- Sodium (Na):≤0.005 %
- Heavy metals (as Pb):≤5 ppm
- Insoluble matter in water:≤0.005 %
- Iodine consuming subst.:Passes test
Frequently Bought Together