3-Aminopyrazine-2-carboxylic acid, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: 3-Amino-2-pyrazinecarboxylic acid
CAS Number:5424-01-1
:
Formula:C₅H₅N₃O₂
MDL Number:MFCD00006141
Molecular Weight:139.11 g/mol
:M-Clarity™ quality level = MQ100
EINECS:226-558-7
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together