Infusion filter, Sterifix® Neonat
BRAU4099257
: B. Braun
- Membrane:0,2 µm
- Colour:Transparent
- Description:Infusion filter with Luer-lock fitting
- Environmentally Preferable:
- Pk:1
Infusion filter with bacteria and particle retention.
- Reliable air venting regardless of position
- High throughput and Supor® membrane
- Pressure resistant to one bar