Specifications
- Description:Miniboom maintenance sorbents
- Environmentally Preferable:
- Pk:6
- Ø×L:76×2400 mm
Specifications
About this item
Chemical sorbent mini booms for laying out around equipment to contain spills and prevent leaks from spreading to neighboring work areas.
The industrial binding fleece consists mainly of inert polypropylene and polyester fibers. The lightweight and dust-free product has a high absorption capacity and reduces disposal costs to a minimum. Mini hoses for targeted containment and absorption of leaks at the base of machines.
- For absorption of oils, coolants and non aggresive liquids
- Booms do not float - absorb water based solutions
- Lightweight and easy to deploy
Recommended Industries include Agriculture, Automotive, Chemicals, Construction, General Manufacturing, Healthcare, Military, Mining, Oil & Gas, Pharmaceuticals, Primary Metal, Transportation
For spill areas in chemical manufacturing, docks/piers/boats, laboratories
For spill types like acids/alkalis/alcohols, chemicals, flammable liquids, solvents - hydrocarbon chlorinated