Order Entry
Poland
ContactUsLinkComponent
Vitamin K1 (phytomenadione, phylloquinone)
Vitamin K1 (phytomenadione, phylloquinone)
Catalog # 47773.
Supplier:  Merck
CAS Number:  
undefined
Vitamin K1 (phytomenadione, phylloquinone)
Catalog # 47773.
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 2-Methyl-3-((7R,11R,E)-3,7,11,15-tetramethylhexadec-2-en-1-yl)naphthalene-1,4-dione
 :
 :450.6958 g/mol
EINECS:201-564-2
CAS Number:84-80-0
 :81818-54-4
Formula:C₃₁H₄₆O₂
MDL Number:MFCD00214063