Calcium acetate 5% in aqueous solution, pure analytical reagent
Supplier: CHEMPUR
Certificates
About this item
Synonyms: Calcium acetate, United States Pharmacopeia (USP) Reference Standard, Calcium di(acetate), Acetic acid calcium salt
CAS Number:62-54-4
:
Formula:(H₃CCOO)₂Ca
MDL Number:MFCD00150019
Molecular Weight:158.17 g/mol
Merck Index:12,01683
Storage Temperature:Ambient
EINECS:200-540-9
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together