Sulphathiazole, Supelco®
Catalog # S9876-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
Specifications
About this item
Synonyms: 4-Amino-N-(thiazol-2-yl)benzenesulphonamide, N1-2-Thiazolylsulphanilamid
:
:255.32 g/mol
EINECS:200-771-5
CAS Number:72-14-0
Formula:C₉H₉N₃O₂S₂
MDL Number:MFCD00005319