Calcium chloride 2,75% (w/v) in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Chloro calcium, Calcium dichloride, CaCl2
CAS Number:10043-52-4
:
Formula:CaCl₂
MDL Number:MFCD00010903
Boiling Pt:1600
Molecular Weight:110.98 g/mol
Melting Pt:772
Merck Index:14,01659
Storage Temperature:Ambient
EINECS:233-140-8
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together