tert-Amyl methyl ether ≥97%, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: TAME, METHOXYPENTANE™, Methyl 2-methyl-2-butyl ether, 1,1-Dimethylpropyl methyl ether, Methyl tert-pentyl ether
CAS Number:994-05-8
:
Formula:C₆H₁₄O
MDL Number:MFCD00010186
Boiling Pt:85…86 °C (lit.)
Density:0.76…0.78 g/mL (25 °C) (lit.)
Molecular Weight:102.17 g/mol
Flash Pt:–7 °C
EINECS:213-611-4
ADR:3 II
:M-Clarity™ quality level = MQ200
UN:3271
:GHS02
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together