Order Entry
Poland
Orders LinkContactUsLinkComponent
Aluminium selenide ≥99% (metals basis)

Aluminium selenide ≥99% (metals basis)

Glass bottle for solids, 1 g
Catalog # 13132.03
Toxic
Health
Environment
Aluminium selenide ≥99% (metals basis)
Catalog # 13132.03
CAS Number:  1302-82-5
Aluminium selenide ≥99% (metals basis)
Aluminium selenide ≥99% (metals basis)

Glass bottle for solids, 1 g

CAS Number:  1302-82-5
Price
707 zł
Log in for availability
1 * 1 g
Product Details & Documents

Purity: 99% CAS No.: 1302-82-5 Molecular Formula: Al2Se3 Formula Weight: 290.84 Physical Form: 6.5mm & Down Pieces Density: 3.437 Appearance: Pale brown Odour: Odourless Solubility: Decomposes to Al203 and SeO2 in water Merck Reference: 14,361 UN No.: 3283 MDL No.: MFCD00014158

Synonyms:  Aluminum selenide
  • CAS Number:1302-82-5
  • Formula:Al₂Se₃
  • MDL Number:MFCD00014158
  • Molecular Weight:290.84 g/mol
  • Merck Index:14,361
  • Melting Pt:960 °C
  • Density:3.437
  • Storage Temperature:Ambient
  • EINECS:215-110-6
  • UN:3283
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.