Sodium hydroxide ≥97%, beads 20-40 mesh, Reagent Grade
Supplier: Honeywell Chemicals
Certificates
About this item
Synonyms: Caustic soda
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
Boiling Pt:1.390 °C (1.013 hPa)
Melting Pt:319 °C
Density:2.13 g/cm³ (20 °C)
Molecular Weight:40 g/mol
Storage Temperature:Ambient
EINECS:215-185-5
ADR:8 II
Merck Index:14,08627
UN:1823
REACH:01-2119457892-27
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Form:beads
- Assay:Min. 96.5 %
- Carbonate (as Na₂CO₃):Max. 2.5 %
- Granulation:complying
- Particle Size:20-40 mesh
- Vapor Pressure:<18 mmHg ( 20 °C)
Specification Test Results
Frequently Bought Together