Benzoin ethyl ether, Sigma-Aldrich®
8.20022.0050
: Merck
:
- Pack type:Glass bottle
- Pk:50 g
- Seq:3368
- Environmentally Preferable:
- Solubility:(20 °C) slightly soluble
- Melting Point:56 - 59 °C
- Molar Mass:240.30 g/mol
- Synonyms:Benzoin ethyl ether, 2-Ethoxy-2-phenylacetophenone
- Hill Formula:C16H16O2
- Chemical Formula:C6H5CHO(C2H5)COC6H5
: Benzoin Ethyl Ether, O-Ethylbenzoin
:
:240.3 g/mol
:209-366-8
:574-09-4
:C₁₆H₁₆O₂
:MFCD00009242
:M-Clarity™ quality level = MQ200