Blood collection tubes, with ACD, BD Vacutainer®
These ACD tubes are used to obtain a whole blood or a plasma sample.
- Contains the active anticoagulant tri-sodium citrate with citric acid and dextrose
- Available in two formulations, ACD solution A and ACD solution B
Tubes are available in glass only and are identifiable by a yellow cap.
Frequently Bought Together