Dropping funnels, cylindrical, with pressure balance, DURAN®
DURAN®, borosilicate glass 3.3, clear, with glass key.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Standard ground joint cone and socket, with divisions for approximate volume reading.
* non-DIN ISO size
: DIN ISO 4800
Frequently Bought Together