n-Heptadecane ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: N-heptadecane
CAS Number:629-78-7
:
Formula:H₃C(CH₂)₁₅CH₃
MDL Number:MFCD00009002
Boiling Pt:302 °C
Melting Pt:22 °C
Density:0.778
Molecular Weight:240.47 g/mol
Flash Pt:148
EINECS:211-108-4
:GHS08
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together