Methanol ≥99.9%, ULTRA RESI-ANALYZED® for gas chromatography, for purge and trap GC analysis, J.T.Baker®
Catalog # 9077-02
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 99.9 %
- Assay (CH₃OH) (by GC, corrected for water):≥99.9 %
- Residue after Evaporation:≤1.0 ppm
- Titrable Acid (µeq/g):≤0.3
- Titrable Base (µeq/g):≤0.10
- Water (by KF, coulometric):≤0.08 %
- Volatile Organic Trace Analysis - Below EPA 8260B CRQL:Conforms / Fails
Specification Test Results
About this item
Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
:
:32.04 g/mol
EINECS:200-659-6
UN:1230
REACH:01-2119433307-44
:GHS02
:GHS06
:GHS08
Storage Temperature:Ambient
CAS Number:67-56-1
Formula:CH₃OH
MDL Number:MFCD00004595
Boiling Pt:64.6
Melting Pt:–98 °C
Density:0.7918 g/cm³ (20 °C)
Flash Pt:11 °C (closed cup)
ADR:3 II
Merck Index:13,05984
:Packaged under inert gas.