Order Entry
Poland
ContactUsLinkComponent
 
1-Pentanol 99%, pure
 
undefined
1-Pentanol 99%, pure
 

 

  • Environmentally Preferable:

 

 

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4070 to 1.4110 (20°C, 589 nm)
  • Water:
    ≤0.2 %
  • GC:
    ≥98.5 %
  • Color scale:
    ≤20 APHA

 

 

Frequently Bought Together