Ammonium acetate, EMSURE® Reag. Ph. Eur., ACS for analysis, Supelco®
Supplier: Merck
Certificates
About this item
Synonyms: Acetic acid ammonium salt
CAS Number:631-61-8
:
Formula:H₃CCOONH₄
MDL Number:MFCD00013066
Density:1.17 g/cm³ (20 °C)
Molecular Weight:77.08 g/mol
Melting Pt:114
Flash Pt:136
Merck Index:13,00495
:M-Clarity™ quality level = MQ300
EINECS:211-162-9
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:1480 g/l (4 °C)
- Melting Point:114 °C
- Molar Mass:77.08 g/mol
- Bulk Density:410 kg/m3
- Flash Point:136 °C
- Density:1.17 g/cm3 (20 °C)
- pH:6.7 - 7.3 (50 g/l, H2O, 25 °C)
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.01116.1000_TDS_EN ProductPage_101116
Product specification
Merck Pharma Segment Brochure
Product Literature
msds_101116
SDS
Frequently Bought Together