2-Heptanone for synthesis, Sigma-Aldrich®
: Merck
:110-43-0
:
:C₇H₁₄O
:MFCD00009513
:150
:–35 °C
:0.8111 g/cm³ (20 °C)
:114.19 g/mol
:43
:203-767-1
:1110
:3 III
:13,04683
:M-Clarity™ quality level = MQ200
:GHS02
:GHS07
:Ambient
- Environmentally Preferable:
- Solubility:4.3 g/l (20 °C)
- Melting Point:-35 °C
- Molar Mass:114.18 g/mol
- Boiling Point:150 - 152 °C (1013 hPa)
- Vapor Pressure:4.5 hPa (20 °C)
- Flash Point:43 °C
- Refractive Index:1.4007 (20 °C, 589 nm)
- Explosion Limit:1.11 - 7.9 %(V)
- Density:0.82 g/cm3 (20 °C)
- Ignition Point:532 °C
- Saturation concentration:16.2 g/m3 (20 °C) Air
- Evaporation number:17.4
- Hazard Class:3
- Hazard Class Description:Flammable Liquid
Frequently Bought Together