1-Bromo-1-chloroethane ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:593-96-4
:
Formula:BrClHCCH₃
MDL Number:MFCD00018842
Boiling Pt:82…83 °C
Density:1.667
Molecular Weight:143.41 g/mol
Storage Temperature:Ambient
EINECS:209-820-5
UN:2810
ADR:6.1 III
:GHS06
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together