(S)-(-)-Limonene 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: (-)-Limonene, (S)-1-Methyl-4-(prop-1-en-2-yl)cyclohex-1-ene, L(-)-Limonene, (S)-(-)-p-Mentha-1,8-diene, (S)-(-)-Dipentene, (S)-(-)-4-Isopropenyl-1-methyl-1-cyclohexene, (S)-(-)-1-Methyl-4-(1-methylvinyl)cyclohexene
CAS Number:5989-54-8
:
Formula:C₁₀H₁₆
MDL Number:MFCD00001558
Boiling Pt:175
Melting Pt:104…84 °C
Density:0.842 g/cm³ (20 °C)
Molecular Weight:136.24 g/mol
Flash Pt:48 °C
UN:2052
ADR:3 III
Merck Index:13,05515
EINECS:227-815-6
:GHS02
:GHS07
:GHS08
:GHS09
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4690 to 1.4760 (20°C, 589 nm)
- GC:≥95 %
- Specific optical rotation:(20°C, 589 nm) (neat) ≤ -60°
Specification Test Results
Frequently Bought Together