Nitric acid 68%, TECHNICAL
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Nitric acid solution
CAS Number:7697-37-2
:
Formula:HNO₃
MDL Number:MFCD00011349
Boiling Pt:120.5 °C (1013 hPa)
Melting Pt:–42 °C
Density:1.4 g/cm³ (20 °C)
Molecular Weight:63.01 g/mol
Storage Temperature:Ambient
EINECS:231-714-2
ADR:8 (5.1) II
Merck Index:13,06608
UN:2031
:GHS03
:GHS05
:GHS06
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:68 - 70 %
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Packaging Information
spec_VWRC20406.292 spec_VWRC20406.320 spec_VWRC20406.320 spec_VWRC20406.361
Product specification
Frequently Bought Together