Ethylamine 70% 70% in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Monoethylamine, Aminoethane, Ethanamine
CAS Number:75-04-7
:
Formula:C₂H₇N
MDL Number:MFCD00008160
Boiling Pt:17
Melting Pt:-81.1
Density:0.8 g/cm³
Molecular Weight:45.08 g/mol
Flash Pt:–17 °C
Storage Temperature:Ambient
UN:2270
ADR:3 II
Merck Index:13,03797
EINECS:200-834-7
:GHS06
:GHS05
:GHS02
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear solution
- Refractive index:1.3830 to 1.3850 (20°C, 589 nm)
- Color scale:≤10 APHA
- Titration with HCl:69 to 71 %
Specification Test Results
Frequently Bought Together