Order Entry
Poland
ContactUsLinkComponent
di-Sodium β-glycerophosphate pentahydrate 98%
di-Sodium β-glycerophosphate pentahydrate 98%
Certificates
undefined
di-Sodium β-glycerophosphate pentahydrate 98%
Certificates
Specifications
  • Environmentally Preferable:
Specifications
Specification Test Results
  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Titration with HClO4:
    ≥97.5 %
  • Isomer:
    ≤2 % alpha-Isomer