Lugol's iodine solution
Catalog # 499319OO
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:50 mL
- Seq:2370
- Environmentally Preferable:
Specifications
About this item
:
Density:1.01 g/cm³ (20 °C)
Storage Temperature:Ambient