Order Entry
Poland
ContactUsLinkComponent
L(-)-Menthol 99.5%
L(-)-Menthol 99.5%
Certificates
undefined
L(-)-Menthol 99.5%
Certificates
Specifications
  • Environmentally Preferable:
Specifications
Specification Test Results
  • Appearance:
    Colorless to white Crystals or crystalline needles
  • Infrared spectrum:
    Conforms
  • GC:
    ≥99.4 %
  • Specific optical rotation:
    -48° to -51° (20°C, 589 nm) (c=10, EtOH)