Digital rotational viscometers, DV-E+
: AMETEK
Digital rotational viscometer, with direct display of viscosity in cP or mPa/s, or % torque, number of revolutions and type of spindle.
- Simplified controls allowing operators to change test parameters quickly
- Digital display for simultaneous measurement of viscosity and torque
- Automatic range to determine full scale range viscosity
Low viscosity (LV) materials include inks, oils, and solvents. Medium viscosity (RV) include creams, food, and paints. High viscosity (HA) materials include gels, chocolate and epoxies. Very high viscosity (HB) materials include asphalt, caulking compounds, and molasses.
: Supplied with case, stand, spindle set and calibration certificate.
: * 1 mPa.s achieved with UL adapter accessory - see later entry, 15 mPa.s on LV with standard spindles
** Minimum viscosity is achieved with optional RV/HA/HB-1 spindle see 616-3202 in entry Accessories
M=1 million; 1 mPa.s= 1 cP
** Minimum viscosity is achieved with optional RV/HA/HB-1 spindle see 616-3202 in entry Accessories
M=1 million; 1 mPa.s= 1 cP
Frequently Bought Together