Bags, zip seal, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Ideal for small lab items, botanical samples and other specimens. Not recommended for storing liquids. LDPE, 101 µm.
- Strong, transparent and waterproof
- Heavy-duty zipper seals contents
Specifications
- Catalog No:
- 216-8121
- 216-8122
- 216-8123
- 216-8124
- 216-8125
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement