1,2,3-Triazole 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 1,2,3-1H-Triazole, 1H-1,2,3-Triazole
CAS Number:288-36-8
:
Formula:C₂H₃N₃
MDL Number:MFCD00014490
Boiling Pt:203 °C (752 mmHg)
Melting Pt:23…25 °C
Density:1.192 g/cm³
Molecular Weight:69.07 g/mol
Flash Pt:107 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4940 to 1.4980 (20°C, 589 nm)
- GC:≥96.0 %
- Titration with HClO4:96.0 to 104.0 %
Specification Test Results
Frequently Bought Together