Sodium chloride 0,85% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Saline, NaCl, Sea salt, Halite
CAS Number:7647-14-5
:
Formula:NaCl
MDL Number:MFCD00003477
Boiling Pt:1413
Molecular Weight:58.44 g/mol
Melting Pt:801
Merck Index:14,08599
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:231-598-3
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together