Methanol-[D₄] (98% D) with 0.03% TMS for NMR spectroscopy
Catalog # 84116.0010
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- Seq:2275
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (on anhydrous substance):Min. 99.9 %
- Isotopic enrichment (FT NMR 400 MHz)(D):Min. 99.80 %
- Water (HDO+D2O):Max. 0.03 %
Specification Test Results
About this item
Synonyms: Methanol-[²H₄], Hydroxy methane-D4, Methanol-d4, Methyl-D3 alcohol-D, Carbinol-D4, Wood alcohol-D4, Methyl alcohol-D4, Tetradeuteromethanol
:
:36.01 g/mol
Flash Pt:11 °C (closed cup)
EINECS:212-378-6
UN:1230
:GHS02
:GHS06
:GHS08
Storage Temperature:Ambient
CAS Number:811-98-3
Formula:D₃COD
MDL Number:MFCD00044637
Boiling Pt:65.3 °C (1013 hPa)
Melting Pt:–99 °C
Density:0.888 g/cm³ (20 °C)
ADR:3 (6.1) II