Anti-PELP1 Rabbit Monoclonal Antibody [clone: ARC1387]
ANTIA305770-100
New Product
- Antibody type:Primary
- Antigen name:glutamic acid- and leucine-rich protein 1
- Antigen symbol:PELP1
- Clonality:Monoclonal
- Clone:ARC1387
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# Q8IZL8
- Antigen synonyms:HMX3|PELP1|PELP1 proline glutamic acid leucine rich protein 1|proline and glutamic acid rich nuclear protein|MNAR|Modulator of nongenomic activity of estrogen receptor|PELP1_HUMAN|Modulator of non-genomic activity of estrogen receptor|P160
- Storage buffer:Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,02% sodium azide
- Molecular weight:160 kDa
- Sequence:EPEPGLLLEVEEPGTEEERGADTAPTLAPEALPSQGEVEREGESPAAGPPPQELVEEEPSAPPTLLEEETEDGSDKVQPPPETPAEEEMETETEAEALQEK
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 1000 - 1100 of human PELP1 (Q8IZL8).
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Rabbit monoclonal [ARC1387] antibody to PELP1 for WB with samples derived from human, mouse and rat.
- Validated applications: WB
- Recommended dilutions: WB: 1:500 to 1:2000
Type: Primary
Antigen: PELP1
Clonality: Monoclonal
Clone: ARC1387
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat