Order Entry
Poland
ContactUsLinkComponent
Dichloromethane-[D₂] (99.8% D) for NMR spectroscopy
Dichloromethane-[D₂] (99.8% D) for NMR spectroscopy
Supplier:  VWR Chemicals
Certificates
undefined
Dichloromethane-[D₂] (99.8% D) for NMR spectroscopy
Supplier:  VWR Chemicals
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 99.9 %
  • Isotopic enrichment (FT NMR 400 MHz)(D):
    Min. 99.80 %
  • Water:
    Max. 0.02 %

Specification Test Results

Frequently Bought Together