1,4-Dibromobenzene-[D₄], Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: 1,4-Dibromobenzene-d4, 1,4-Dibromobenzene-[²H₄]
CAS Number:4165-56-4
:
Formula:C₆D₄Cl₂
MDL Number:MFCD00045021
Boiling Pt:219…221 °C (1013 hPa)
Melting Pt:88…90 °C
Density:1.8 g/cm³ (20 °C)
Molecular Weight:239.87 g/mol
Storage Temperature:Ambient
UN:3077
ADR:9 III
:M-Clarity™ quality level = MQ100
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together