2-Iodobenzoyl chloride, Sigma-Aldrich®
Catalog # SIAL252115-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
About this item
Synonyms: 2-Iodobenzoylchloride, o-Iodobenzoyl chloride
:
:266.47 g/mol
EINECS:210-196-1
UN:3261
:GHS05
Storage Temperature:Ambient
CAS Number:609-67-6
Formula:C₇H₄ClIO
MDL Number:MFCD00001040
Boiling Pt:290
Melting Pt:27…30 °C
ADR:8 II