Order Entry
Poland
ContactUsLinkComponent
Anti-scFv Rabbit Monoclonal Antibody Cocktail (Biotinylated)
Anti-scFv Rabbit Monoclonal Antibody Cocktail (Biotinylated)
Catalog # GENSA02283-200
Supplier:  Genscript
Anti-scFv Rabbit Monoclonal Antibody Cocktail (Biotinylated)
Catalog # GENSA02283-200
Supplier:  Genscript

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    scFv
  • Clonality:
    Cocktail
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG kappa
  • Reactivity:
  • Environmentally Preferable:
  • Form:
    Liquid
  • Antigen synonyms:
    Single-chain variable region fragment|Single-chain fv|VH and VL of an antibody|VH-linker-VL|VL-linker-VH|Single-chain fragment variable|Nanobody|Single chain antibody
  • Storage buffer:
    Supplied in PBS (pH 7,2), containing 10 mg/ml BSA, 0,02% sodium azide and 50% glycerol
  • Storage temperature:
    Store at ‒20 °C
  • Concentration:
    0,5 mg/ml
  • Purification:
    Protein A affinity column
  • Pk:
    200 µl

Specifications

About this item

This product is specific for scFvs in different species (humanised, mouse) and different orders (VH-linker-VL, VL-linker-VH). It can also recognise various forms of scFv-based bispecific antibodies, such as DART, BITE, TandAbs, IgG(H)-scFv2 and scFv-Fab-Fc.

A single chain fragment variable (scFv) is a type of recombinant antibody. It is approximately 25 kDa and consists of the variable regions of the heavy (VH) and light (VL) chains of an antibody, which are connected by a flexible peptide linker. scFvs have several advantages, including their small molecular weight, strong penetration, and high specificity. They play crucial roles in targeted therapy, imaging diagnosis, and biological detection. Importantly, in the field of cell therapy, scFvs can act as the antigen recognition domain of CAR-T cells and determine the targeting ability of these cells.

Caution: For research use only. Not intended for human and animal therapeutic or diagnostic use.

Type: Primary
Antigen: scFv
Clonality: Monoclonal
Clone: Cocktail
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG, κ
Reactivity: Human, Mouse