4-Bromo-N-methylaniline 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6911-87-1
:
Formula:C₇H₈BrN
MDL Number:MFCD03030391
Boiling Pt:123…125 °C (7 mmHg)
Molecular Weight:186.05 g/mol
UN:2810
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow to brown to dark purple Liquid
- Infrared spectrum:Conforms
- Refractive index:1.6110 to 1.6150 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together