Sterile Cleanroom Face Masks with Tyvek® Side Extensions
Supplier: KEYSTONE CLEANROOM PRODUCTS
Certificates
About this item
Sterile cleanroom face masks feature Tyvek® side extensions to prevent skin exposure. Double elastic headband fastening makes it easier to put on compared to ties. Optional fog-free design prevents goggles from fogging and mask slippage.
- Tyvek® side extensions provide wider coverage area and an improved fit, while also eliminating exposed skin
- Optional adhesive nose strip prevents goggles from fogging
- Innovative design prevents mask slippage, reducing contamination risks and significant material and labour costs associated with re-gowning
- Prevents the common observations noted by inspectors during audits related to facial skin exposure inside aseptic processing areas
- Packaged in chevron clean-peel Tyvek® pouches
Aseptic operators are more effective and productive when they can see clearly and are comfortable.
Certifications: Certificates of Sterility and Conformance.
Packaging: 25 facemasks per chevron clean-peel Tyvek® pouch. Each package is labelled with a lot number for traceability and an expiration date. Each Tyvek® pouch is contained in a sealed poly bag.
Frequently Bought Together